Magnetic induction stirrers
Supplier: HEATHROW
120584 120585
HEAT120584EA 1270 SGD
HEAT120584 HEAT120585
Magnetic induction stirrers
Stirrers Magnetic Stirrers
The magnetic induction stirrer is motor-less, using advanced coil technology that produces the strongest magnetic coupling and has no moving parts making this unit 100% wear-free and maintenance free.
  • Easy to read digital display precisely controls time and speed enabling reproducible conditions
  • Unit is programmable with 50% duty up to 99 cycles
  • Time and speed can be conveniently adjusted while the unit is running
  • Reverse rotation option for better mixing results
  • Speed remains constant, even when load changes
  • Rapid acceleration and braking (less than 10
More Product Information