Cell culture media, RPMI 1640
Supplier: PAN-BIOTECH GMBH
P04-17500
P04-18000
P04-16516
P04-18047
P04-17550
P03-4310
P03-4350
P03-7310
P03-7350
P04-16151
P04-16500
P04-16515
P04-16520
P04-16598
P04-17510
P04-17545
P04-17546
P04-17553
P04-17598
P04-17599
P04-17850
P04-18500
P04-19500
P04-19550
P04-21049
P04-22100
P04-22500
P03-4301
P04-16100
P03-7301
P04-22550
P04-16555
P04-17597
P04-02601
P04-18043
P04-18057
P04-18598
P04-17345
P04-17556
P04-18058
P03-76610
PANBP04-17500EA
141
SGD
PANBP04-17500
PANBP04-18000
PANBP04-16516
PANBP04-18047
PANBP04-17550
PANBP03-4310
PANBP03-4350
PANBP03-7310
PANBP03-7350
PANBP04-16151
PANBP04-16500
PANBP04-16515
PANBP04-16520
PANBP04-16598
PANBP04-17510
PANBP04-17545
PANBP04-17546
PANBP04-17553
PANBP04-17598
PANBP04-17599
PANBP04-17850
PANBP04-18500
PANBP04-19500
PANBP04-19550
PANBP04-21049
PANBP04-22100
PANBP04-22500
PANBP03-4301
PANBP04-16100
PANBP03-7301
PANBP04-22550
PANBP04-16555
PANBP04-17597
PANBP04-02601
PANBP04-18043
PANBP04-18057
PANBP04-18598
PANBP04-17345
PANBP04-17556
PANBP04-18058
PANBP03-76610
Cell culture media, RPMI 1640
Cell Culture Media
RPMI 1640 medium was developed for culture of normal and neoplastic leukocytes, but also marrow cells and hybridoma cells.
- Serum-free media for hybridoma cells
Supplementing RPMI 1640 with varying amounts of FBS, a very good medium for many different cell lines can be obtained.