Protein-free media, Panserin S2
Supplier: PAN-BIOTECH GMBH
P04-710200 P04-710210
PANBP04-710200EA 0 SGD
PANBP04-710200 PANBP04-710210
Protein-free media, Panserin S2
Cell Culture Media
Panserin S2 is a protein-free medium for an optimised growth of insect Drosophila S2 cells in suspension culture. Insect cells are widely used for the industrial production of recombinant proteins.
  • Store at 2 to 8 °C
It contains amino acids, vitamins, salts, trace elements, lipids and growth promoting factors in a formulation optimised for the growth of insect cells. It contains no protein or any further components of human or animal origin.
It is suitable for the cultivation of Drosophila S2 cells and the production of recombinant protein. (e.g. Baculovirus expression vector system, BEVS).
With its protein-free formulation is free of human and animal components. This allows the production of recombinant proteins for medical and therapeutic purposes. The protein-free formulation also facilitates convenient and economic purification of final products from the cell culture. Panserin S2 guarantees a high cell density and viability resulting in an increased production and easy and economic purification of recombinant protein. (Baculovirus expression vector system).
Caution: For research purpose only. Not suitable for human or animal diagnostic or therapeutics procedures.