Cell culture media, Endopan PRO
Supplier: PAN BIOTECH
P04-0070K P04-00700 P04-0070S
PANBP04-0070KEA 0 SGD
PANBP04-0070K PANBP04-00700 PANBP04-0070S
Cell culture media, Endopan PRO
Cell Culture Media
Endopan PRO ready-to-use is a complete medium specially developed for the in vitro culture of human progenitor endothelial cells (hPEC) containing all components necessary for optimal colony formation, clonogenic growth, and rapid proliferation.
  • Sterile medium
  • Store at ‒20 °C
Endopan PRO Kit is provided with FBS growth supplement (pre-screened and tested for progenitor cells) and additional supplements in separate sterile packing. This will enable the user to prepare a medium for special applications.
Caution: For research use only. Not approved for human or animal diagnostic or therapeutic procedures.