Cell culture supplements, Panexin basic, serum replacement
Supplier: PAN-BIOTECH GMBH
P04-96900 P04-96090K
PANBP04-96900EA 396 SGD
PANBP04-96900 PANBP04-96090K
Cell culture supplements, Panexin basic, serum replacement
Cell Culture Supplements and Additives
Panexin basic is a fully defined serum replacement for the cultivation of adherent and non-adherent cells under serum-free culture conditions or to significantly reduce the amount of serum in cell culture. It supports the growth of many cell types in an optimum manner without any extra handling compared to serum.
  • Sterile solution
  • Store at ‒20 °C
Panexin basic contains purified proteins, lipids, salts, amino acids, trace elements, hormones and a 3-dimensional substrate release system in an optimised formulation. It contains no growth factors, undefined hydrolysates or peptones.
Panexin basic is designed to replace or to reduce serum in the cell culture in a very simple manner. In most cases there is no need to change the basal medium. As Panexin basic is fully defined and contains no peptones or hydrolysates, lot testing is no more necessary. It also allows high reproducibility and a simplified downstream process.
Panexin basic contains no growth factors and enables defined proliferation and differentiation of stem cells. Characterisation studies of growth factors will obtain more reproducible and clearer results. Panexin basic is also useful to develop sensitive cell-based in vitro tests and coculture procedures.
Caution: For Research use only.