Bovine serum albumin (BSA), Fraction V
Supplier: PAN-BIOTECH GMBH
P06-1395100 P06-1395500
PANBP06-1395100EA 0 SGD
PANBP06-1395100 PANBP06-1395500
Bovine serum albumin (BSA), Fraction V
Proteins and Peptides
Albumins serve as additive proteins for tissue cultures. They are the main protein component in serum and are added to cell culture media to increase the stability of cell membranes and to bind possibly toxic elements.
  • Minimal lot to lot variation
  • Stringent control of raw materials
  • Minimal BSE/TSE risk
  • US origin
  • Full documentation
  • Special products for unique applications