Human serum albumin
Supplier: PAN-BIOTECH GMBH
P06-26025 P06-26050 P06-26105 P06-27100 P06-26110 P06-26150 P06-27500 P06-260100 P06-261100
PANBP06-26025EA 2840 SGD
PANBP06-26025 PANBP06-26050 PANBP06-26105 PANBP06-27100 PANBP06-26110 PANBP06-26150 PANBP06-27500 PANBP06-260100 PANBP06-261100
Human serum albumin
Sera (Serum)
Human serum albumin is the most abundant protein in blood. Albumin transports hormones, fatty acids and other components.
  • Absebce of chemical and microbial contamination
  • Manifactured under conditions certified to ISO 9001
  • Serves as additive protein for tissue cultures
  • Increase the stability of cell membranes
The main biological function is to regulate the colloidal osmotic pressure of blood. Human serum albumin can be used for protein calibration studies and solubilization of lipids.
Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV½ and Anti-HCV and nonreactive for HBsAg, HIV-1 Antigen and RPR with kits approved by the FDA.
Due to their free hydrophobic region, fatty acid free albumin is used to solubilise lipids in tissue culture. It is also used as a blocking agent in Western blots or ELISA technique and is also an effective cryoprotectant.