Cell culture supplements, Neuropan 2

Supplier: PAN-BIOTECH GMBH

P07-11010 P07-11100 P07-11005 P07-11050
PANBP07-11010EA 1540 SGD
PANBP07-11010 PANBP07-11100 PANBP07-11005 PANBP07-11050
Cell culture supplements, Neuropan 2
Cell Culture Supplements and Additives

Neuropan-2 supplement is a chemically-defined, serum-free supplement for the growth of primary embryonic neurons as well as embryonic neurospheres/CNS progenitor cells when combined with bFGF and EGF in Neuropan basal medium.


  • Formulated to meet special requirements of neuronal cells
  • Sterile solution
  • Store at ‒20 °C
  • Contains insulin, transferrin, albumin and hormones


When supplemented with Neuropan basal medium it allows for long-term maintenance of the normal phenotype and growth of neuronal cells, and maintain pure populations of neuronal cells without the need for an astrocyte feeder.


Neuropan-2 supplement meets the special cell culture requirements of pre-natal and embryonic neuronal cells from hippocampus, cortex and other regions of the brain.


Caution: For research use only. Not approved for human or animal diagnostic or therapeutic procedures.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR