Cell culture media, Marrowpan
Supplier: PAN BIOTECH
P04-70200 P04-70250
PANBP04-70200EA 0 SGD
PANBP04-70200 PANBP04-70250
Cell culture media, Marrowpan
Cell Culture Media
Marrowpan is a complete medium intended for in vitro diagnostic procedures with a short term culture of bone marrow and other hematopoietic cells for cytogenetic studies. It is intended for in vitro use and has been designed for establishing cultures of bone marrow and leukemic blood cells, which then can be used in karyotyping, fluorescence in-situ hybridisation (FISH) or other cytogenetic procedures.
  • Ready-to-use
  • Suitable for the cultivation of cells from peripheral blood or bone marrow
  • Store at –20 °C
It is based on alpha-MEM and contains antibiotics, L-glutamine, fetal bovine serum (FBS), hormones and growth factors.
It can be used as a neutral medium to culture different haematopoietic cells (myeloid and lymphoid lineages) present in bone marrow or leukemic blood samples. Marrowpan is also used together with a mitogen specific for B or T lymphocytes where these particular lineages are being investigated.
It is suitable for a rapid expansion of blood cells in order to investigate leukemic diseases (e.g. ALL, AML, CLL, CML, MPN, MDS). The number and quality of metaphases in Marrowpan are significantly higher and independent of individual batches as compared to serum-containing media.
Caution: For research purpose only. Not suitable for human or animal diagnostic or therapeutics procedures.