Cell culture media, Amniopan III
Supplier: PAN-BIOTECH GMBH
P04-70103 P04-70503
PANBP04-70103EA 0 SGD
PANBP04-70103 PANBP04-70503
Cell culture media, Amniopan III
Cell Culture Media
Amniopan III is a complete ready-to-use medium intended for in vitro diagnostic use with a short term culture of human fetal cells from amniotic fluid or chorion villi biopsy (CVS) material for a standardised application in cytogenetic studies.
  • Ready-to-use
  • Store at –20 °C
It is intended for in vitro use and has been designed for establishing cultures of human fetal cells from amniotic fluid or chorion villi biopsies (CVS), which then can be used in karyotyping, fluorescence in-situ hybridisation (FISH) or other cytogenetic procedures.
Amniopan III is newly developed formulation is optimal to facilitaterapid diagnostic findings.
Caution: For research purpose only. Not suitable for human or animal diagnostic or therapeutics procedures.