Cell culture media, Medium 199
Supplier: PAN-BIOTECH GMBH
P04-07050 P04-07150 P04-07250 P04-07450 P04-07500 P04-07600 P04-07753 P03-2150 P03-1910 P03-1950 P03-2110 P03-1901 P03-2101 P04-07561 P04-07053 P04-07050SA1
PANBP04-07050EA 202 SGD
PANBP04-07050 PANBP04-07150 PANBP04-07250 PANBP04-07450 PANBP04-07500 PANBP04-07600 PANBP04-07753 PANBP03-2150 PANBP03-1910 PANBP03-1950 PANBP03-2110 PANBP03-1901 PANBP03-2101 PANBP04-07561 PANBP04-07053 PANBP04-07050SA1
Cell culture media, Medium 199
Cell Culture Media
The M199 was originally developed to assay the nutrient demand of embryonic chicken fibroblasts. But it works very well with cells from many different animal species. For example, it is used for vaccine production in virology. For long term cultures serum should be added.
  • Store from 2 to 8 °C