Kanamycin sulphate 50 mg/ml
Supplier: PAN-BIOTECH GMBH
|
Danger
|
Synonyms:
Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
Even though cells should be cultured without the addition of antibiotics or antifungal drugs, cell culture media are not only a perfect breeding ground for your cell culture but also for many contaminants, such as bacteria and fungi. In certain cases the addition of antibiotics is necessary (for unique cell lines or to remove already existing contamination).
Formula:
C₁₈H₃₈N₄O₁₅S MW: 582,6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...