Cell culture media, SILAC-Dulbecco's modified eagle medium
Supplier: PAN BIOTECH
P04-02506 P04-02514 P04-02509 P04-02551 P03-02515 P03-02506
PANBP04-02506EA 0 SGD
PANBP04-02506 PANBP04-02514 PANBP04-02509 PANBP04-02551 PANBP03-02515 PANBP03-02506
Cell culture media, SILAC-Dulbecco's modified eagle medium
Cell Culture Media
DMEM for SILAC is optimised for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyse protein expression by mass spectrometry (MS).
  • Liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
  • High quality, medium is sterile, endotoxin-free, and cell culture-compatible