Cell culture media, CMRL 1066
Supplier: PAN BIOTECH
P04-84500 P04-84600 P04-84700 P03-8410
PANBP04-84500EA 0 SGD
PANBP04-84500 PANBP04-84600 PANBP04-84700 PANBP03-8410
Cell culture media, CMRL 1066
Cell Culture Media
CMRL – 1066 is a chemically defined medium, which was developed in the late 1950’s by Connaught Medical Research Laboratories. In the past it was developed to clone monkey-kidney cells and as a long time culture medium for L-cells.
  • An nucleoside and vitamin-rich medium
  • Designed for unsupplemented culture of L-strain-cells
  • Used to maintain monolayer growth of permanent cell lines for years
Suitable for many types of human and monkey cells and also for other mammalian cells, especially by using horse and calf serum.