Serum-free media, Panserin 801
Supplier: PAN BIOTECH
P04-710801 P04-710801B P04-710801K
PANBP04-710801EA 1420 SGD
PANBP04-710801 PANBP04-710801B PANBP04-710801K
Serum-free media, Panserin 801
Cell Culture Media
Panserin 801 is a serum-free, ready-to-use medium for the cultivation of human keratinocytes.
  • Store at 2 to 8 °C
It has been developed for the serum-free cultivation of human keratinocytes. It selectively supports the growth of human keratinocytes and concurrently prevents the overgrowth with fibroblasts.