Cell culture media, Grace's insect media
Supplier: PAN-BIOTECH GMBH
P03-9010
P03-9050
P03-9110
P03-9150
P04-81500
P04-82500
P04-82520
PANBP03-9010EA
877
SGD
PANBP03-9010
PANBP03-9050
PANBP03-9110
PANBP03-9150
PANBP04-81500
PANBP04-82500
PANBP04-82520
Cell culture media, Grace's insect media
Cell Culture Media
Grace's insect medium is used to culture cells derived from a variety of insects including several species of lepidopterans, as well as some dipterans, for growth and maintenance of cell lines.
- Sterile-filtered
- Supports a broad range of lepidopteran cells
The Grace's insect medium was originally developed to culture insect cells including SF9 and SF21 cells.