Foetal bovine serum, FBS Dialysed, South American origin
Supplier: PAN-BIOTECH GMBH
P30-2101 P30-2102
PANBP30-2101EA 0 SGD
PANBP30-2101 PANBP30-2102
Foetal bovine serum, FBS Dialysed, South American origin
Sera (Serum)
FBS dialysed is a specially processed serum product. Serum is ultrafiltrated against 0,15 M NaCl solution with a 10000 Da MW cutoff filter.
  • Reproducible growth properties
  • Performance tested
  • Ideal for radiolabeling assays
  • Suitable for a great variety of cells
  • Continuous quality control
  • 0,2 µm sterile filtered
  • Store at ‒20 °C
This ultrafiltration process is carefully designed to significantly reduce the concentrations of small molecules such as amino acids hormones or cytokines. FBS dialysed is used in applications requiring serum depleted of these before mentioned small molecular weight molecules, that are necessary for alternative biochemical survival pathways. The glucose level, which is representative of the extent of dialysis, is below 10 mg/dl after completion of dialysis.