Filtrated bovine serum, FBS Good Forte, Australia origin
Supplier: PAN-BIOTECH GMBH
P40-49100 P40-49500
PANBP40-49100EA 1160 SGD
PANBP40-49100 PANBP40-49500
Filtrated bovine serum, FBS Good Forte, Australia origin
Sera (Serum)
FBS Good Forte contains serum and additional growth fortifying compounds to increase cell proliferation. In addition, growth promoting and stabilising compounds (e.g. proteins, salts, sugars, vitamins) have been added to further enhance the stability of the serum as well as the proliferation of many different cell types.
  • Reproducible enhanced growth properties
  • Very low endotoxin level
  • Continued high quality
  • No more batch testing required
  • Store at ‒20 °C
  • 0,2 µm sterile filtered