Foetal bovine serum biotech, Australia origin
Supplier: PAN BIOTECH
P40-1301 P40-1302
PANBP40-1301EA 0 SGD
PANBP40-1301 PANBP40-1302
Foetal bovine serum biotech, Australia origin
Sera (Serum)
Bovine serum is the blood fraction remaining after the natural coagulation of blood, followed by centrifugation to remove any remaining red blood cells. Fetal bovine serum (FBS) containing an especially high amount of growth factors is the most widely used serum.
  • Sterile filtered 0,2 µm
  • Stored at −20 °C
The production of bovine serum at is tightly controlled, from the collection of serum at the slaughterhouse and throughout the whole production cycle which is performed without exception in own production facilities.
Certifications: Tested according to EMEA CPMP/BWP/1793/02 and Ph. Eur. 01/2008:2262.