Cell culture media, Earl's balanced salts (EBBS)
Supplier: PAN BIOTECH
P04-30010P
P04-30050P
P04-30500
P04-31500
P04-38500
P04-39500
P04-47500
P04-40100
P04-43500
PANBP04-30010PEA
1310
SGD
PANBP04-30010P
PANBP04-30050P
PANBP04-30500
PANBP04-31500
PANBP04-38500
PANBP04-39500
PANBP04-47500
PANBP04-40100
PANBP04-43500
Cell culture media, Earl's balanced salts (EBBS)
Cell Culture Media
The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions. These solutions are usually used for washing tissues and cells. Often they are combined with other agents in cell and tissue treatment.
- Store from 2 °C to room temperature