Cell culture media, Hank's balanced salts (HBBS)
Supplier: PAN-BIOTECH GMBH
P04-34500 P04-32010P P04-32050P P04-32100 P04-32105 P04-32500 P04-32505 P04-33010P P04-33100 P04-34100 P04-49100 P04-49105 P04-49500 P04-50100 P04-50105 P04-50500 P04-50505 P04-49200 P04-49300
PANBP04-34500EA 200 SGD
PANBP04-34500 PANBP04-32010P PANBP04-32050P PANBP04-32100 PANBP04-32105 PANBP04-32500 PANBP04-32505 PANBP04-33010P PANBP04-33100 PANBP04-34100 PANBP04-49100 PANBP04-49105 PANBP04-49500 PANBP04-50100 PANBP04-50105 PANBP04-50500 PANBP04-50505 PANBP04-49200 PANBP04-49300
Cell culture media, Hank's balanced salts (HBBS)
Cell Culture Media
The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions. These solutions are usually used for washing tissues and cells. Often they are combined with other agents in cell and tissue treatment.
  • Store from 2 °C to room temperature