(±)-1-Phenyl-1-pentanol 95%

Supplier: Thermo Scientific
Warning

Synonyms: Fenipentol

L04646.09 L04646.18
ALFAL04646.09EA 58.2 SGD
ALFAL04646.09 ALFAL04646.18
(±)-1-Phenyl-1-pentanol 95%
(±)-1-Phenyl-1-pentanol

Purity: 95% CAS No.: 583-03-9 Molecular Formula: C11H16O Formula Weight: 164.25 Refractive Index: 1.5080 Boiling Point: 137°/2mm Merck Reference: 14,3975 MDL No.: MFCD00021935

Formula: C₆H₅CH(OH)(CH₂)₃CH₃
MW: 164,25 g/mol
Boiling Pt: 137 °C (2 mmHg)
Density: 0,960
Storage Temperature: Ambient
MDL Number: MFCD00021935
CAS Number: 583-03-9
EINECS: 209-493-9
Merck Index: 14,3975

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR