Kanamycin sulphate 5 mg/ml
Supplier: PAN BIOTECH
|
Danger
|
P06-04001
P06-04050
P06-04100
P06-04025
PANBP06-04001EA
0
SGD
PANBP06-04001
PANBP06-04050
PANBP06-04100
PANBP06-04025
Kanamycin sulphate 5 mg/ml
Kanamycin sulphate
Even though cells should be cultured without the addition of antibiotics or antifungal drugs, cell culture media are not only a perfect breeding ground for your cell culture but also for many contaminants, such as bacteria and fungi. In certain cases the addition of antibiotics is necessary (for unique cell lines or to remove already existincontamination).
Formula:
C₁₈H₃₈N₄O₁₅S Storage Temperature: Freezer |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...