Cell culture media, Waymouth's MB 752/1
Supplier: PAN-BIOTECH GMBH
P04-28500
P03-4510
P03-4550
PANBP04-28500EA
875
SGD
PANBP04-28500
PANBP03-4510
PANBP03-4550
Cell culture media, Waymouth's MB 752/1
Cell Culture Media
Waymouth's is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism, and growth of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment.
- Used for growing strain L sublines, NCTC clone 929
The Waymouth's MB 752/1 media was developed for studies concerning nutrition and metabolism.