Serum-free media, Panserin 401
Supplier: PAN-BIOTECH GMBH
P04-710401
P04-710401S2
PANBP04-710401EA
440
SGD
PANBP04-710401
PANBP04-710401S2
Serum-free media, Panserin 401
Cell Culture Media
Panserin 401 is a fully defined complete ready-to-use medium for the serum-free cultivation of a multitude of adherent and non adherent cells.
- Multi-purpose medium suitable for a variety of cells
- Store at 2 to 8 °C
Based on Iscove's MEM, trace elements, albumin, cholesterol, soya lipids and vitamins were added to the medium. It does not contain any growth or attachment factors.
As the medium contains no growth factors there is a possibility to investigate the effects of specific growth factors added to the cell culture.