Gentamicin sulphate 10 mg/ml

Supplier: PAN-BIOTECH GMBH
Warning

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

P06-03001 P06-03050 P06-03100 P06-03025
PANBP06-03001EA 1490 SGD
PANBP06-03001 PANBP06-03050 PANBP06-03100 PANBP06-03025
Gentamicin sulphate 10 mg/ml
Gentamicin sulphate

Even though cells should be cultured without the addition of antibiotics or antifungal drugs, cell culture media are not only a perfect breeding ground for your cell culture but also for many contaminants, such as bacteria and fungi. In certain cases the addition of antibiotics is necessary (for unique cell lines or to remove already existing contamination).

Storage Temperature: Freezer
MDL Number: MFCD00270181
CAS Number: 1405-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR