Cell culture media, Schneider's Drosophila medium
Supplier: PAN BIOTECH
P03-9310 P03-9350 P03-9410 P03-9450 P04-90500 P04-91500 P04-90599
PANBP03-9310EA 930 SGD
PANBP03-9310 PANBP03-9350 PANBP03-9410 PANBP03-9450 PANBP04-90500 PANBP04-91500 PANBP04-90599
Cell culture media, Schneider's Drosophila medium
Cell Culture Media
Schneider's Drosophila medium was developed for the culture of Drosophila cells.
  • Sterile media
  • Store at 2 to 8 °C
Also suitable for the culture of other dipteran cell lines.