Foetal bovine serum, Sera Low, South American origin
Supplier: PAN BIOTECH
P30-5100 P30-5500
PANBP30-5100EA 0 SGD
PANBP30-5100 PANBP30-5500
Foetal bovine serum, Sera Low, South American origin
Sera (Serum)
Sera Low is a specially selected serum with low endotoxin level. The endotoxin level is lower than 1 EU/ml.
  • Higher safety (viruses, mycoplasma)
  • Suitable for a great variety of cells
  • Permanent quality control
  • 0,2 µm sterile filtered
  • Store at ‒20 °C