Cell culture media, Ham's F10
Supplier: PAN-BIOTECH GMBH
P03-3910 P03-3950 P03-4010 P03-4050 P03-5010 P03-5050 P04-12049 P04-12050 P04-13050 P04-13500 P04-12100 P04-12010
PANBP03-3910EA 793 SGD
PANBP03-3910 PANBP03-3950 PANBP03-4010 PANBP03-4050 PANBP03-5010 PANBP03-5050 PANBP04-12049 PANBP04-12050 PANBP04-13050 PANBP04-13500 PANBP04-12100 PANBP04-12010
Cell culture media, Ham's F10
Cell Culture Media
Ham's F10 is an alternative to Ham's F12 and it was used primarily to culture CHO-cells. Ham's F10 can be used with or without FBS for many different cell cultures. It is used for example for primary cells of rat and chicken, but also for human diploid cells.
  • Sterile
  • Store from 2 to 8 °C