Cell culture media, Liebovitz L-15 medium
Supplier: PAN-BIOTECH GMBH
P04-27050 P04-27054 P04-27055 P04-27500 P03-1510 P03-1550 P03-1610 P03-1650 P03-16110
PANBP04-27050EA 243 SGD
PANBP04-27050 PANBP04-27054 PANBP04-27055 PANBP04-27500 PANBP03-1510 PANBP03-1550 PANBP03-1610 PANBP03-1650 PANBP03-16110
Cell culture media, Liebovitz L-15 medium
Cell Culture Media
Liebovitz L-15 contains no sodium hydrogen carbonate because it is buffered already by a high concentration of amino acids.
  • Store at 2 to 8 °C
It supports the growth of established cells like Hep-2, but also human nerve cells and primary tissue explants. With 10 % tryptose phosphate broth it is also ideally suited for the cultivation of insect cell lines.