Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

NP015-10 NP015-20
BJAANP015-10EA 2790 SGD
BJAANP015-10 BJAANP015-20
Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Acetonitrile

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃CCN
Boiling Pt: 82 °C (1,013 hPa)
Melting Pt: -46 °C
Density: 0,7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=9f9a68a160195b7256f30345f34dbda929c419dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
Fluorescence (as quinine base) 0.3 ppb max.
GC Analysis 99.9 % min.
HPLC Gradient @ 215 nm 0.005 AU max.
HPLC Gradient @ 254 nm 0.001 AU max.
Refractive Index (20ºC) 1.3434 - 1.3446
Residue 1 mg/L max.
UV Cutoff 190 nm max.
Water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR