Human platelet lysate
Supplier: PAN BIOTECH
P40-29005 P40-29020 P40-29050 P40-29100
PANBP40-29005EA 0 SGD
PANBP40-29005 PANBP40-29020 PANBP40-29050 PANBP40-29100
Human platelet lysate
Cells
Platelets are a natural source of growth factors they contain growth factors (platelet-derived growth factors (PDGFs), basic ibroblast growth factor (bFGF), transforming growth factor (TGF-β) and insulin-like growth factor-1 (IGF-1).
  • Lead to a strong proliferation of human cells in culture
  • Able to promote Mesenchymal stem cells (MSC) expansion
  • Decrease the time required to reach conluence
  • Increase CFU-F size, as compared to the FBS medium
  • Promotes the expansion of human adherent and non-adherent primary cells and cell lines
Human Platelet Lysate (hPL) is a xeno-free alternative to FBS. Human Platelet Lysate is a well promising source of bioactive substances as growth factors not only for invivo wound healing and tissue repair, but also for the expansion of human cells in culture.
Human platelet lysate is a powerful and safe substitute for development of tissue- and cell-engineered products using MSCs.
The single sample collections are taken from human, healthy donors in certiied donation institutions in Europe. he collection of blood follows the EU standards (Directive 2002/98/EC - quality and safety standards for the collection, testing, processing, storage and distribution of human blood and blood components).
According to the EU standards the donors were tested for the following infection parameters: HBsAg, HBV DNA, anti-HCV, HCV RNA, anti-HIV, HIV RNA and Lues serology. It is possible to exclude the window period in case of NAT and antibodies / antigen tests (e.g. HBsAG and HBV NAT) with the current state-of-the-art test according to EU guidelines.
Caution: Heparin is not included in this product. Heparin is necessary to reduce the precipitaion caused by fibrinogen.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions -20 °C