n-Heptane ≥99%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

Synonyms: high purity solvents

NP207-20 207-1
BJAANP207-20EA 3740 SGD
BJAANP207-20 BJAA207-1
n-Heptane ≥99%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
n-Heptane

For HPLC, GC, pesticide residue analysis, and spectrophotometry. UV cutoff at 200nm.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 °C
Melting Pt: –90.6 °C
Density: 0.684 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis (n-Heptane) 99 % min.
GC Analysis (toal C7 isomers) 99.9 % min.
Refractive Index (20ºC) 1.3872 - 1.3884
Residue 1 mg/L max.
UV Absorbance @ 225 nm 0.1 AU max.
UV Absorbance @ 250 nm 0.01 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Cutoff 200 nm max.
Water by Karl Fischer Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR