3-Chloro-4-iodoaniline ≥98%
Supplier: Thermo Scientific
|
Danger
|
A14256.06
A14256.14
A14256.22
ALFAA14256.06EA
0
SGD
ALFAA14256.06
ALFAA14256.14
ALFAA14256.22
3-Chloro-4-iodoaniline ≥98%
3-Chloro-4-iodoaniline
Purity: 98%
CAS No.: 135050-44-1
Molecular Formula: C6H5ClIN
Formula Weight: 253.47
Melting Point: 65-71°
Beilstein Reference: 3236464
UN No.: 2811
MDL No.: MFCD00051845
Formula:
C₆H₅ClIN MW: 253,47 g/mol Boiling Pt: 289 °C (1013 hPa) Melting Pt: 65…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051845 CAS Number: 135050-44-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...