5-Chloro-2-iodoaniline ≥98%

Supplier: Thermo Scientific
Danger

B25709.03 B25709.06
ALFAB25709.03EA 0 SGD
ALFAB25709.03 ALFAB25709.06
5-Chloro-2-iodoaniline ≥98%
5-Chloro-2-iodoaniline

Purity: 98% CAS No.: 6828-35-9 Molecular Formula: C6H5ClIN Formula Weight: 253.47 Melting Point: 40-42° UN No.: 2811 MDL No.: MFCD06738960

Formula: C₆H₅ClIN
MW: 253,47 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD06738960
CAS Number: 6828-35-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR