ω-Bromo-4-cyanoacetophenone ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone

B25382.06 B25382.14
ALFAB25382.06EA 104 SGD
ALFAB25382.06 ALFAB25382.14
ω-Bromo-4-cyanoacetophenone ≥97%
ω-Bromo-4-cyanoacetophenone

Purity: 97% CAS No.: 20099-89-2 Molecular Formula: C9H6BrNO Formula Weight: 224.06 Odour: Irritating Melting Point: 91-96° Beilstein Reference: 2087648 UN No.: 2923 MDL No.: MFCD00052931

Formula: C₉H₆BrNO
MW: 224,06 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00052931
CAS Number: 20099-89-2
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR