dNTP mix

Supplier: PAN-BIOTECH GMBH

PAN739028 PAN739043 PAN739029
PANBPAN739028EA 1220 SGD
PANBPAN739028 PANBPAN739043 PANBPAN739029
dNTP mix
Nucleic Acid Reagents Nucleotides

A ready-to-use molecular grade dNTP Mix containing dATP, dCTP, dGTP and dTTP at pH 7,5 as lithium salts in purified
water. The mix is designed to save hands-on time for researchers and minimise the possibility of contamination.


  • Convenient, pre-optimised and pre-mixed
  • Ultra-pure: >99% trisphosphate by HPLC
  • Extended shelf-life of 24 months at –20 °C
  • Free from PCR inhibitors
  • DNase, RNase, protease, phosphatase and nickase free


Applications: standard and long range PCR assays, cDNA synthesis, qPCR, microarrays, DNA sequencing, DHPLC, labelling.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR