5-Iodo-o-toluidine ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Amino-4-iodotoluene, 5-Iodo-2-methylaniline

L07404.03 L07404.06
ALFAL07404.03EA 0 SGD
ALFAL07404.03 ALFAL07404.06
5-Iodo-o-toluidine ≥98%
5-Iodo-o-toluidine

Purity: 98% CAS No.: 83863-33-6 Molecular Formula: C7H8IN Formula Weight: 233.05 Melting Point: 46-49° Beilstein Reference: 2078769 UN No.: 2811 MDL No.: MFCD00051531

Formula: C₇H₈IN
MW: 233,05 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00051531
CAS Number: 83863-33-6
EINECS: 281-094-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR