Tetrafluoro-p-benzoquinone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

L01287.03 L01287.06
ALFAL01287.03EA 96.7 SGD
ALFAL01287.03 ALFAL01287.06
Tetrafluoro-p-benzoquinone ≥97%
Tetrafluoro-p-benzoquinone

Purity: 97% CAS No.: 527-21-9 Molecular Formula: C6F4O2 Formula Weight: 180.06 Melting Point: ca 184° subl. Beilstein Reference: 1875039

Formula: C₆F₄O₂
MW: 180,06 g/mol
Boiling Pt: 133 °C (1013 hPa)
Melting Pt: ∼ 184 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00001592
CAS Number: 527-21-9
EINECS: 208-411-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR