Barium titanate (∼35 wt% Oxide) polymeric precursor

Supplier: Thermo Scientific
Warning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

39755.04 39755.09 39755.18
ALFA39755.04EA 111 SGD
ALFA39755.04 ALFA39755.09 ALFA39755.18
Barium titanate (∼35 wt% Oxide) polymeric precursor
Barium titanate

Common Applications: Used in piezoelectric devices.
Notes: Resulting product is BaTiO3. Crystallization temperature is 650 °C
Soluble in alcohols.


CAS No.: 12047-27-7 Physical Form: Viscous Liquid Solubility: Soluble in alcohols UN No.: 1993 MDL No.: MFCD00003447


Used in piezoelectric devices

Formula: BaO₃Ti
MW: 233,21 g/mol
Melting Pt: 1625 °C
Density: 5,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1993
ADR: 3,III
Merck Index: 12,01029

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR