Barium titanate ≥99.9% (metals basis)
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
88266.14
88266.22
ALFA88266.14EA
213
SGD
ALFA88266.14
ALFA88266.22
Barium titanate ≥99.9% (metals basis)
Barium titanate
Purity: 99.9%
CAS No.: 12047-27-7
Molecular Formula: BaTiO3
Formula Weight: 233.19
Physical Form: 3-12mm Pieces
Density: 5.85
Melting Point: 1620°
Merck Reference: 14,1000
MDL No.: MFCD00003447
For vacuum deposition
Formula:
BaO₃Ti MW: 233,21 g/mol Melting Pt: 1620 °C Density: 5,85 Storage Temperature: Ambient |
MDL Number:
MFCD00003447 CAS Number: 12047-27-7 EINECS: 234-975-0 UN: 1564 ADR: 6.1,III Merck Index: 14,1000 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...