1,5-Bis(diphenylphosphino)pentane ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane

B21054.03 B21054.06 B21054.14
ALFAB21054.03EA 0 SGD
ALFAB21054.03 ALFAB21054.06 ALFAB21054.14
1,5-Bis(diphenylphosphino)pentane ≥97%
1,5-Bis(diphenylphosphino)pentane

Purity: 97% CAS No.: 27721-02-4 Molecular Formula: (C6H5)2P(CH2)5P(C6H5)2 Formula Weight: 440.50 Melting Point: 46-48° MDL No.: MFCD00003052

Formula: C₂₉H₃₀P₂
MW: 440,5 g/mol
Boiling Pt: 230 °C (0,1 mmHg)
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003052
CAS Number: 27721-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR