1,6-Dimethoxynaphthalene ≥98%
Supplier: Thermo Scientific
Synonyms:
2,5-Dimethoxynaphthalene
L18129.06
L18129.03
ALFAL18129.06EA
329
SGD
ALFAL18129.06
ALFAL18129.03
1,6-Dimethoxynaphthalene ≥98%
1,6-Dimethoxynaphthalene
Purity: 98+%
CAS No.: 3900-49-0
Molecular Formula: C12H12O2
Formula Weight: 188.23
Melting Point: 58-59°
Boiling Point: 123°/0.8mm
MDL No.: MFCD00086330
Formula:
C₁₂H₁₂O₂ MW: 188,23 g/mol Boiling Pt: 123 °C (0,8 mmHg) Melting Pt: 58…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00086330 CAS Number: 3900-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...