Human recombinant BMP-4 (from E. coli)

Supplier: PAN BIOTECH

P-3610002 P-3610003
PANBP-3610002EA 0 SGD
PANBP-3610002 PANBP-3610003
Human recombinant BMP-4 (from E. coli)
Proteins and Peptides

Bone morphogenetic protein 4 (BMP-4) human recombinant produced in E. coli is a monomeric, nonglycosylated, polypeptide chain containing 116 amino acids having a molecular mass of 13 kDa. It is purified by proprietary chromatographic techniques.


Sequence: SPKHHSQRAR KKNKNCRRHS LYVDFSDVGW NDWIVAPPGY QAFYC HGDCP FPLADHLNST NHAIVQTLVN SVNSSIPKAC CVPTELSAIS MLYLDEYDKV VLKNYQEMV EGCGCR


Reconstitute the lyophilised BMP-4 in sterile ddH₂O not less than 100 µg/ml which can then be further diluted to other aqueous solutions.


Caution: For research use only. Not approved for human or animal diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name BMP-4
Environmentally Preferable
Protein synonyms ZYME|BMP4|BMP2B
Protein/peptide type Recombinant
Species Human
Source E. coli
Purity >95% as determined by RP-HPLC and SDS-PAGE
Formulation Lyophilised from a 0,2 µm filtered concentrated (1 mg/ml) solution in 20 mM Na₂CO₃ buffer, pH 9,0
Molecular weight 13 kDa
Storage conditions –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR