Serum-free media, PowerStem MSC1
Supplier: PAN-BIOTECH GMBH
PowerStem MSC1 is an easy to use xeno-free medium without animal derived components (xeno-free) for cultivation and proliferation of human mesenchymal stem cells (hMSC). It is especially designed for the proliferation of human mesenchymal stem cells without differentiation. It supports longterm growth of MSC and preserves their multi-lineage potential.
- Store at –20 °C
It contains salts, amino acids, trace elements, hormones, growth factors, and enriched human proteins and lipids in an optimised formulation. It is free of animal derived components (xeno-free).