Transfection reagent, PANFect
Supplier: PAN BIOTECH
P02-8000K P02-8010 P02-8110 P02-8210 P02-8020 P02-8050 P02-8010K P02-8110K P02-8210K
PANBP02-8000KEA 0 SGD
PANBP02-8000K PANBP02-8010 PANBP02-8110 PANBP02-8210 PANBP02-8020 PANBP02-8050 PANBP02-8010K PANBP02-8110K PANBP02-8210K
Transfection reagent, PANFect
Transfection Reagents
PANFect is a multipurpose transfection reagent for pDNA, siRNA and mRNA delivery as well as cotransfection into a broad range of cell lines as HEK, HeLa, Mef, adherent cells, cells in suspension for iPS and also primary cell lines.
  • Easy and fast optimisation protocol
  • Very gentle transfection reagent with low toxicity
  • Free of animal derived components
  • Serum compatible and no need to change media
  • Suitable for standard transfections, genome editing and high throughput screenings
PanFect A shows low cytotoxicity and highly efficient for common cell lines and suitable for difficult transfect cells such as mouse embryonic cells (mESC).
PanFect A-plus is a multipurpose transfection reagent with optimised formulation requires less reagents per transaction. Suitable for pDNA, siRNA, and CRISPR/Cas.
PanFect siRNA is suitable for the delivery of small interfering RNA duplexes for gene lockdown. Suitable for siRNA, and miRNA.
Delivery information: Kits include reagents, booster and buffer solutions.