Chemical resistant overalls, MICROCHEM® 4000 AVANT AIRline, Model 755
Supplier: Ansell
GR40T-00755-02 GR40T-00755-06 GR40T-00755-05 GR40T-00755-04 GR40T-00755-03 GR40T-00755-07
ANSEGR40T-00755-02EA 1770 SGD
ANSEGR40T-00755-02 ANSEGR40T-00755-06 ANSEGR40T-00755-05 ANSEGR40T-00755-04 ANSEGR40T-00755-03 ANSEGR40T-00755-07
Chemical resistant overalls, MICROCHEM® 4000 AVANT AIRline, Model 755
Disposable Garments
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Exceptional protection; over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, offering the highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves
  • Emergency rip cord enables rapid suit removal in cases of emergency or undue distress
Fully encapsulated suit designedMore Product Information
Order Now
SPECIFICATIONS
Model 755
Environmentally Preferable
Gender
Garment application Chemical protection