2-Acetyl-3-methylpyrazine ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(3-Methylpyrazinyl)ethan-1-one, 1-(3-Methylpyrazin-2-yl)ethanone

A15054.03 A15054.14
ALFAA15054.03EA 0 SGD
ALFAA15054.03 ALFAA15054.14
2-Acetyl-3-methylpyrazine ≥98%
2-Acetyl-3-methylpyrazine

Purity: 98% CAS No.: 23787-80-6 Molecular Formula: C7H8N2O Formula Weight: 136.15 Flash Point: 100°(212°F) Refractive Index: 1.5220 Boiling Point: 265-266° Beilstein Reference: 742438 MDL No.: MFCD00014612

Formula: C₇H₈N₂O
MW: 136,15 g/mol
Boiling Pt: 265…266 °C
Density: 1,105
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014612
CAS Number: 23787-80-6
EINECS: 245-889-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR