2-Acetylpyrazine ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Pyrazin-2-ylethan-1-one, Acetylpyrazine
A15917.03
A15917.06
ALFAA15917.03EA
0
SGD
ALFAA15917.03
ALFAA15917.06
2-Acetylpyrazine ≥99%
2-Acetylpyrazine
Purity: 99%
CAS No.: 22047-25-2
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Melting Point: 75-78°
Boiling Point: 78-79°/8mm
MDL No.: MFCD00006134
Formula:
C₆H₆N₂O MW: 122,13 g/mol Boiling Pt: 78…79 °C (8 mmHg) Melting Pt: 75…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006134 CAS Number: 22047-25-2 EINECS: 244-753-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...