2-Bromo-4-nitropyridine-N-oxide ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Bromo-4-nitropyridine N-oxide, 2-Bromo-4-nitropyridine 1-oxide
L20020.03
L20020.06
ALFAL20020.03EA
160
SGD
ALFAL20020.03
ALFAL20020.06
2-Bromo-4-nitropyridine-N-oxide ≥97%
2-Bromo-4-nitropyridine N-oxide
Purity: 97%
CAS No.: 52092-43-0
Molecular Formula: C5H3BrN2O3
Formula Weight: 219.00
Melting Point: 143-146°
Beilstein Reference: 145698
Formula:
C₅H₃BrN₂O₃ MW: 218,99 g/mol Melting Pt: 143…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00160743 CAS Number: 52092-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...